missing translation for 'onlineSavingsMsg'
Learn More

Glucagon R/GCGR Antibody (hGR-2 F6), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18368900 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368900 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368900 Supplier Novus Biologicals Supplier No. NBP281065AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glucagon R/GCGR Monoclonal specifically detects Glucagon R/GCGR in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucagon R/GCGR
Applications Immunofluorescence
Classification Monoclonal
Clone hGR-2 F6
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ97182, GGR, GL-R, glucagon receptor, MGC138246
Gene Symbols GCGR
Host Species Mouse
Immunogen Recombinant human Glucagon R/GCGR.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2642
Test Specificity The antibody binds to the human glucagon receptor, Glucagon R/GCGR.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.