missing translation for 'onlineSavingsMsg'
Learn More

Glucagon R/GCGR Antibody (591929) [DyLight 650], Novus Biologicals™

Product Code. 18539345 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18539345 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18539345 Supplier Novus Biologicals Supplier No. FAB10296W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glucagon R/GCGR Monoclonal antibody specifically detects Glucagon R/GCGR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucagon R/GCGR
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 591929
Conjugate DyLight 650
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ97182, GGR, GL-R, glucagon receptor, MGC138246
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Glucagon R/GCGR, Met1-Gln142, Accession # P47871
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2642
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.