missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucagon Antibody (53), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05184AF647
This item is not returnable.
View return policy
Description
Glucagon Monoclonal specifically detects Glucagon in Mammal samples. It is validated for Western Blot, ELISA.
Specifications
| Glucagon | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GCG | |
| Synthetic GLP-1(9-18) adsorbed onto aluminum hydroxide gel | |
| 0.1 mL | |
| Cancer, GPCR, Signal Transduction | |
| 2641 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 53 | |
| Western Blot, ELISA | |
| glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Based on inhibition ELISA, It cross-reacts <2% with GLP-1(7-36)amide | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction