missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glucagon (19-29), Human

Product Code. p-5384763
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 15415689

Brand: Thermo Scientific Chemicals J63443.M^

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cellular and molecular biology applications

This fragment inhibits calcium activated ATPase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Format Powder
For Use With (Application) This fragment inhibits calcium activated ATPase activity
Molecular Weight (g/mol) 1352.50
Product Type Glucagon (19-29), human
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.