missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glucagon (1-29) trifluoroacetate salt, human

Product Code. 15403539
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403539 0.5 mg 0.5mg
15413539 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15403539 Supplier Thermo Scientific Chemicals Supplier No. J60827.!D
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cellular and molecular biology applications

A peptide hormone that plays a role in maintaining glucose homeostasis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Format Powder
For Use With (Application) A peptide hormone that plays a role in maintaining glucose homeostasis
Molecular Weight (g/mol) 3482.74
Product Type Glucagon (1-29), human
Quantity 0.5 mg
Regulatory Status RUO - research use only

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.