missing translation for 'onlineSavingsMsg'
Learn More

Glubionate Calcium Hydrate, TRC

Product Code. 30395876
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395876 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395876 Supplier Toronto Research Chemicals Supplier No. TRCG412750500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium Glubionate
CAS 12569-38-9
Molecular Formula C12 H21 O12 . C6 H11 O7 . Ca . H2 O
Quantity 500 mg
Synonym Calcium, (4-O-β-D-galactopyranosyl-D-gluconato)(D-gluconato)-, monohydrate, D-Gluconic acid, 4-O-β-D-galactopyranosyl-, calcium complex, D-Gluconic acid, calcium complex, Calcium glubionate, Calcium glucono-galacto-gluconate, Calcium gluconolactobionate
InChI Formula InChI=1S/C12H22O12.C6H12O7.Ca.H2O/c13-1-3(15)10(7(18)8(19)11(21)22)24-12-9(20)6(17)5(16)4(2-14)23-12;7-1-2(8)3(9)4(10)5(11)6(12)13;;/h3-10,12-20H,1-2H2,(H,21,22);2-5,7-11H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t3-,4-,5+,6+,7-,8-,9-,10-,12+;2-,3-,4+,5-;;/m11../s1
SMILES O.[Ca+2].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)[O-].OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(=O)[O-]
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate;(2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanoate;hydrate
Molecular Weight (g/mol) 610.529
Formula Weight 610.127
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.