missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLU1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA598343
Additional Details : Weight : 0.01000kg
Description
Involved in glutamate biosynthesis in leaf. Required for the reassimilation of ammonium ions generated during photorespiration. [Uniprot]Specifications
GLU1 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein G | |
RUO | |
Plant | |
Antibody | |
IgG |
ELISA, Western Blot | |
2.05 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
Recombinant Arabidopsis thaliana Ferredoxin-dependent glutamate synthase 1, chloroplastic/mitochondrial protein (106-505AA) | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |