missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLTPD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566759
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VRSLGARSGCG PRAQPCVPGE TAPFQVRQES GTLE Highest antigen sequence identity to the following orthologs - mouse 37%, rat 37%.
GLTPD2 (Glycolipid Transfer Protein Domain Containing 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include glycolipid binding and glycolipid transporter activity.Specifications
GLTPD2 | |
Polyclonal | |
Unconjugated | |
GLTPD2 | |
C730027E14Rik; Gltpd2; glycolipid transfer protein domain containing 2; glycolipid transfer protein domain-containing protein 2; RGD1560459 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388323 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NH11 | |
GLTPD2 | |
Recombinant Human GLTPD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only