missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLTP Antibody (OTI2A10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45875
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
GLTP Monoclonal specifically detects GLTP in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GLTP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:400 - 1:4000 | |
| glycolipid transfer protein | |
| Mouse | |
| 23.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI2A10 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| GLTP | |
| Full length human recombinant protein of human GLTP(NP_057517) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| 51228 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction