Learn More
Invitrogen™ GLT8D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113919
Description
Immunogen sequence: IRHLGWNPDA RYSEHFLQEA KLLHWNGRHK PWDFPSVHND LWESWFVPDP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 77%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.
Specifications
| GLT8D2 | |
| Polyclonal | |
| Unconjugated | |
| GLT8D2 | |
| 1110021D20Rik; GALA4A; GLT8D2; glycosyltransferase 8 domain containing 2; glycosyltransferase 8 domain-containing protein 2; gycosyltransferase; RGD1560432; UNQ1901/PRO4347 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 83468 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H1C3 | |
| GLT8D2 | |
| Synthetic peptide directed towards the C terminal region of human GLT8D2 (aa 290-339). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.