missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLT8D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532140
This item is not returnable.
View return policy
Description
Recommended positive controls: mouse muscle, Rat muscle. Predicted reactivity: Mouse (92%), Rat (93%), Rhesus Monkey (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the glycosyltransferase family. The specific function of this protein has not been determined. Alternative splicing results in multiple transcript variants of this gene [provided by RefSeq, May 2013].
Specifications
| GLT8D1 | |
| Polyclonal | |
| Unconjugated | |
| Glt8d1 | |
| 2410004H05Rik; 5430414N14Rik; AD-017; AI450005; Da2-24; GALA4A; Glt8d1; glycosyltransferase 8 domain containing 1; glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017; MSTP137; MSTP139; UNQ572/PRO1134 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 306253, 55830, 76485 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q68CQ7, Q6AYF6, Q6NSU3 | |
| Glt8d1 | |
| Recombinant protein encompassing a sequence within the center region of human GLT8D1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction