missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLT6D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554795
This item is not returnable.
View return policy
Description
Immunogen sequence: PLVAQLHAWW YFRNTKNFPY ERRPTSAACI PFGQGDFYYG NLMVGGTPHN ILDFIKEYLN GVIHDIKNGL NSTYE Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 67%.
Specifications
| GLT6D1 | |
| Polyclonal | |
| Unconjugated | |
| GLT6D1 | |
| galactosyltransferase family 6 domain containing 1; galactosyltransferase family 6 domain-containing 1; GLT6D1; GLTDC1; glycosyltransferase 6 domain containing 1; glycosyltransferase 6 domain-containing protein 1; GT6M7; Putative glycosyltransferase 6 domain-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 360203 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z4J2 | |
| GLT6D1 | |
| Recombinant protein corresponding to Human GLT6D1. Recombinant protein control fragment (Product #RP-93105). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction