Learn More
GLT25D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16768-1-AP-20UL
 View more versions of this product
Product Code. 16836504
  • £135.00 / 20µL
Specifications
GLT25D1
Polyclonal
Unconjugated
COLGALT1
GLT25D1
Rabbit
Antigen Affinity Chromatography
RUO
234407, 290637, 79709
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8K297, Q8NBJ5
Colgalt1
GLT25D1 Fusion Protein Ag10346
20 μL
Primary
Rat, Mouse, Human