missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524595
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat based on sequence homology.
The protein encoded by this gene is a mitochondrial phosphate-activated glutaminase that catalyzes the hydrolysis of glutamine to stoichiometric amounts of glutamate and ammonia. This protein is functionally similar to the kidney glutaminase but is a little smaller in size. Originally thought to be liver-specific, this protein has been found in other tissues as well. At least one transcribed pseudogene has been found for this gene.
Specifications
| GLS2 | |
| Polyclonal | |
| Unconjugated | |
| Gls2 | |
| A330074B06Rik; AI195532; breast cell glutaminase; Ga; GLS; GLS2; glutaminase 2; glutaminase 2 (liver, mitochondrial); glutaminase GA; glutaminase I; glutaminase liver isoform, mitochondrial; hLGA; Kiaa4146; L hL L-glutaminase; L hLGA; LGA; L-glutaminase; L-glutamine amidohydrolase; liver mitochondrial glutaminase; phosphate-activated glutaminase; phosphate-dependent glutaminase | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 216456, 27165 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q571F8, Q9UI32 | |
| Gls2 | |
| KLH conjugated synthetic peptide between 498-524 amino acids of human GLS2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction