Learn More
GLS2 Antibody (CL7721) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88908UV
Product Code. 30307817
  • £558.00 / 0.10mL
Description
GLS2 Monoclonal antibody specifically detects GLS2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GLS2
Monoclonal
DyLight 350
EC 3.5.1.2, GAbreast cell glutaminase, GLSglutaminase GA, glutaminase 2 (liver, mitochondrial), glutaminase I, glutaminase liver isoform, mitochondrial, hLGA, LGA, L-glutaminase, L-glutamine amidohydrolase, MGC71567, phosphate-activated glutaminase, phosphate-dependent glutaminase, truncated glutaminase 2
This antibody was developed using a synthetic peptide derived from Q9UI32, with the exact immunogen sequence remaining proprietary.
0.1 mL
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
27165
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL7721
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified