missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLS2 Antibody (CL7716) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88907PE
This item is not returnable.
View return policy
Description
GLS2 Monoclonal antibody specifically detects GLS2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| GLS2 | |
| Monoclonal | |
| PE | |
| EC 3.5.1.2, GAbreast cell glutaminase, GLSglutaminase GA, glutaminase 2 (liver, mitochondrial), glutaminase I, glutaminase liver isoform, mitochondrial, hLGA, LGA, L-glutaminase, L-glutamine amidohydrolase, MGC71567, phosphate-activated glutaminase, phosphate-dependent glutaminase, truncated glutaminase 2 | |
| This antibody was developed using a synthetic peptide derived from Q9UI32, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 27165 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL7716 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction