missing translation for 'onlineSavingsMsg'
Learn More

GLS2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18331110 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331110 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331110 Supplier Novus Biologicals Supplier No. NBP176544AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GLS2 Polyclonal specifically detects GLS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLS2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.5.1.2, GAbreast cell glutaminase, GLSglutaminase GA, glutaminase 2 (liver, mitochondrial), glutaminase I, glutaminase liver isoform, mitochondrial, hLGA, LGA, L-glutaminase, L-glutamine amidohydrolase, MGC71567, phosphate-activated glutaminase, phosphate-dependent glutaminase, truncated glutaminase 2
Gene Symbols GLS2
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid peptide near the center terminus of human GLS2 (NP_037399).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specificity for GLS2 versus GLS1 has not been determined.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.