missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLRX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13381-1-AP-150UL
This item is not returnable.
View return policy
Description
Glutaredoxin (Grx), also known as thiol transferase, is a small heat-stable oxidoreductase. Grxs form part of the glutaredoxin system, comprising NADPH, GSH and glutathione reductase, which transfers electrons from NADPH to glutaredoxins via GSH. First discovered in E. coli as GSH-dependent hydrogen donors for ribonucleotide reductase, Grx catalyzes GSH-disulfide oxidoreductase via two redox-active cysteine residues. The active sequence (Cys-Pro-Tyr-Cys) is conserved in a variety of species. The 12-kDa dithiol protein has a role in reduction of mixed disulfides in cells exposed to oxidative stress.Specifications
| GLRX2 | |
| Polyclonal | |
| Unconjugated | |
| GLRX2 | |
| GLRX2, Glutaredoxin 2, Glutaredoxin 2, mitochondrial, GRX2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114022, 51022, 69367 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AXW1, Q923X4, Q9NS18 | |
| GLRX2 | |
| GLRX2 Fusion Protein Ag4218 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction