missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLRB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562593
This item is not returnable.
View return policy
Description
Immunogen sequence: MLNNPKRVEA EKARIAKAEQ ADGKGGNVAK KNTVNGTGTP VHISTLQVGE TRCKKVCTSK SDLRSNDFSI VGSLPRDFEL SNYDCYGKPI EVNNGLGKSQ AKNNKKPPPA KPVIPTAAK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
In the central nervous system (CNS), glycine-mediated inhibitory neurotransmission is essential to voluntary motor control and reflex responses. Glycine binds to glycine receptors (GlyR) in the postsynaptic neuronal membranes. GlyR, gamma-aminobutryic acid, serotonin and acetylcholine comprise an evolutionally conserved superfamily of ligand-gated ion channels. The pentameric subunit structure of GlyR consists of two types of glycosylated membrane proteins, alpha1 through alpha4 and beta, and an associated peripheral membrane protein, which combine to form a chloride-selective ion channel. In humans, the composition of the pentamer changes from alpha2 subunits in the fetal CNS to alpha1 and beta subunits in the adult CNS. Fast potentiation of GlyR by intracellular Ca2+ in the brainstem and midbrain indicate an important role for Ca2+ in modulation of glycinergic synapses.
Specifications
| GLRB | |
| Polyclonal | |
| Unconjugated | |
| GLRB | |
| AI853901; Glrb; glycine receptor; Glycine receptor 58 kDa subunit; glycine receptor beta; glycine receptor subunit beta; glycine receptor, beta; glycine receptor, beta subunit; GlyR beta; Glyrb; HKPX2; inhibitory glycine receptor; spa; spastic | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2743 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48167 | |
| GLRB | |
| Recombinant protein corresponding to Human GLRB. Recombinant protein control fragment (Product #RP-91139). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction