Learn More
Invitrogen™ GLRA1/GLRA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA184890
Description
PA1-84890 detects a band at ~48 kDa in rat spinal cord lysates. Reconstitute with 0.1 mL PBS. For long term storage, the addition of 0.09% sodium azide is recommended. Prior to reconstitution, store at 4ºC. After reconstitution, store undiluted at -20ºC, avoiding freeze/thaw cycles. Rabbit anti glycine receptor recognizes an epitope within the N-terminal region (NT) of glycine receptor alpha 1 and alpha 2 subunits.
Glycine receptors are members of the ligand-gated ion channel superfamily, which mediate fast inhibitory neurotransmission. The receptors are pentameric membrane proteins which form chloride channels. Binding of glycine to its receptor produces an increase in chloride conductance and membrane hyperpolarisation. Four genes encoding glycine receptor alpha subunits have been identified, together with a single beta polypeptide. Each subunit consists of a large extracellular N-terminal region, four transmembrane domains, and a large cytoplasmic domain.
Specifications
| GLRA1/GLRA2 | |
| Polyclonal | |
| Ammonium bicarbonate with no preservative | |
| P07727, P22771, P23415, P23416, Q64018, Q7TNC8 | |
| GLRA1, GLRA2 | |
| Synthetic peptide corresponding to the N-terminal region of rat glycine receptor alpha 1 subunit. | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| GLRA2 | |
| GLR; Glra1; GLRA2; Glycine receptor 48 kDa subunit; glycine receptor alpha 1; glycine receptor alpha 2; glycine receptor alpha 2 subunit; Glycine receptor alpha 2 subunit (glycine receptor, neonatal); glycine receptor strychnine-binding subunit; Glycine receptor subunit alpha-1; glycine receptor subunit alpha-2; glycine receptor, alpha 1; glycine receptor, alpha 1 subunit; glycine receptor, alpha 2; glycine receptor, alpha 2 subunit; glycine receptor, alpha-2 polypeptide; Glyr; GLYRA1; HKPX1; nmf11; oscillator; ot; RNNEOGLY; spasmodic; spd; STHE | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14654, 237213, 24397, 25674, 2741, 2742 | |
| 4°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.