missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLPG1837, MedChemExpress
Description
GLPG1837 is a potent and reversible CFTR potentiator, with EC50s of 3 nM and 339 nM for F508del and G551D CFTR, respectively.
Specifications
Specifications
| Chemical Name or Material | GLPG1837 |
| Physical Form | Solid |
| CAS | 1654725-02-6 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C16H20N4O3S |
| Synonym | ABBV-974 |
| Solubility Information | DMSO : ≥ 250 mg/mL (717.52 mM) |
| SMILES | O=C(C1=NNC=C1)NC(S2)=C(C(N)=O)C3=C2C(C)(C)OC(C)(C)C3 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?