missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLP2R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576959
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The GLP2 receptor (GLP2R) is a G protein-coupled receptor superfamily member closely related to the glucagon receptor ans GLP1 receptor. Glucagon-like peptide-2 (GLP2) is a 33-amino acid proglucagon-derived peptide produced by intestinal enteroendocrine cells. Like glucagon-like peptide-1 (GLP1) and glucagon itself, it is derived from the proglucagon peptide encoded by the GCG gene. GLP2 stimulates intestinal growth and upregulates villus height in the small intestine, concomitant with increased crypt cell proliferation and decreased enterocyte apoptosis. Moreover, GLP2 prevents intestinal hypoplasia resulting from total parenteral nutrition. GLP2R, a G protein-coupled receptor superfamily member is expressed in the gut and closely related to the glucagon receptor (GCGR) and the receptor for GLP1 (GLP1R).
Specifications
| GLP2R | |
| Polyclonal | |
| Unconjugated | |
| GLP2R | |
| 9530092J08Rik; G protein-coupled receptor GLP2R; GLP-2; Glp2 receptor; GLP-2 receptor; GLP2R; GLP-2R; GLP-2-R; glucagon like peptide 2 receptor; glucagon-like peptide 2 receptor; glucagon-like peptide-2 receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9340, 93896 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O95838, Q5IXF8 | |
| GLP2R | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 460-553 of human GLP2R. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction