Learn More
Invitrogen™ GLP-1 Monoclonal Antibody (53)

Mouse Monoclonal Antibody
Brand: Invitrogen™ ABS0445302
Description
Specificity: ABS 044-53-02 binds human GLP-1 (9-36) amide. Based on inhibition ELISA, this antibody shows less than 2% cross-Specificity with GLP-1 (7-36) amide. Reactivity: ABS 044-53-02 binds free GLP-1 (9-36) amide in solution. This product has a higher affinity for GLP-1 (9-36) amide than ABS 044-49-02 as determined by an Inhibition assay. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
GCG is also known as GLP1, or Glucagon. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis.It is mapped to 2q36-2q37. GLP1, also known as 7-37 for the codons of the preproglucagon molecule which encode it, renders pancreatic beta-cells 'glucose-competent' and may be useful in the treatment of noninsulin-dependent diabetes mellitus. Also, GLP1 is a potent insulin secretagogue.It plays a major role in the enteroinsular axis, accounting, for example, for the finding that plasma insulin levels accompanying oral intake of glucose are greater than those observed when glucose is given intravenously.
Specifications
| GLP-1 | |
| Monoclonal | |
| 1.15 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P01275 | |
| GCG | |
| Synthetic GLP-1(9-18) adsorbed onto aluminum hydroxide gel. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 53 | |
| Unconjugated | |
| GCG | |
| GCG; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; glucagon; Glucagon like peptide 1; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; Incretin hormone; MGC138331; OXM; OXY; Oxyntomodulin; PPG; preproglucagon | |
| Mouse | |
| Protein A | |
| RUO | |
| 2641 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.