missing translation for 'onlineSavingsMsg'
Learn More

GLP1 Antibody (4), DyLight 650, Novus Biologicals™

Product Code. 18413676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413676 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413676 Supplier Novus Biologicals Supplier No. NBP223558C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GLP1 Monoclonal antibody specifically detects GLP1 in Human, Mouse, Mammalia samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLP1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 4
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Host Species Mouse
Immunogen Synthetic GLP-1(7-17)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2641
Target Species Human, Mouse, Mammalia
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.