missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-rp1/IGFBP-7 Antibody (40012B) [mFluor Violet 610 SE], Novus Biologicals™
Goat Monoclonal Antibody
Brand: Novus Biologicals FAB13341MFV610
This item is not returnable.
View return policy
Description
IGFBP-rp1/IGFBP-7 Monoclonal antibody specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Immunohistochemistry
Specifications
| IGFBP-rp1/IGFBP-7 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-rp1, Arg98-Arg277, Accession # AAA16187 | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 3490 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry | |
| 40012B | |
| 50mM Sodium Borate | |
| Goat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction