missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP/EHMT1 Antibody (1H2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079813-M04
This item is not returnable.
View return policy
Description
GLP/EHMT1 Monoclonal antibody specifically detects GLP/EHMT1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| GLP/EHMT1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 1H3-K9-HMTase 5, Eu-HMTase1DEL9q34, EUHMTASE1FP13812, FLJ12879, G9a-like protein 1, GLP, GLP1, H3 lysine-9 specific 5, KIAA1876euchromatic histone methyltransferase 1, KMT1DDKFZp667M072 | |
| EHMT1 (NP_079033, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAADEGSAEKQAGEAHMAADGETNGSCENSDASSHANAAKHTQDSARVNPQDGTNTLTRIAENGVSERDSEAAKQNHVTADDFVQTSVIGSNGYILNKPA | |
| 0.1 mg | |
| Breast Cancer | |
| 79813 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 1H2 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| NP_079033 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction