missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP-1R Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97308H
This item is not returnable.
View return policy
Description
GLP-1R Polyclonal specifically detects GLP-1R in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot, Knockdown Validated.
Specifications
| GLP-1R | |
| Polyclonal | |
| Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot, Knockdown Validated | |
| P43220 | |
| GLP1R | |
| A synthetic peptide made to an internal portion of the human GLP1R protein (between residues 250-350) [UniProt P43220] | |
| 0.1 mL | |
| Cancer, GPCR, Lipid and Metabolism, Signal Transduction | |
| 2740 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot, KnockDown | |
| HRP | |
| PBS with No Preservative | |
| GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction