missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GLP-1 (7-37)

Product Code. 18794709
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18794709 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18794709 Supplier Tocris Bioscience™ Supplier No. 5374/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Endogenous bioactive GLP-1 receptor ligand

GLP-1 (7-37) is a endogenous GLP-1 receptor ligand; bioactive and truncated form of GLP-1; insulinotropic hormone.
TRUSTED_SUSTAINABILITY

Specifications

Purity 0.95
CAS 106612-94-6
Content And Storage Store at -20°C
Description Glucagon-Like Peptide 1 Receptor Agonists
Molecular Weight (g/mol) Observed M.W.: 3355.71
Quantity 1 mg
Structural Form C151H228N40O47
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.