missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress GLP-1(7-36), amide acetate

Product Code. 16469338
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469338 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469338 Supplier MedChemExpress Supplier No. HYP0054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GLP-1(7-36), amide acetate is a major intestinal hormone that stimulates glucose-induced insulin secretion from β cells.

Specifications

Purity 98.29%
CAS 1119517-19-9
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
For Use With (Application) Metabolism-sugar/lipid metabolism
Molecular Weight (g/mol) 3357.73 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (29.78 mM∣Need ultrasonic)
Synonym Glucagon-like peptide-1 (GLP-1)(7-36), amide acetate∣Human GLP-1 (7-36), amide acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.