Learn More
Marigold™ Gloves G25G Industrial Protective
Brand:  Marigold™ 03275
Product Code. 12931105
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
EN420 dexterity grade 5, cotton flock lined, open cuff, raised pattern nitrile EN388 abrasion grade 4, cut grade 1, tear grade 0, puncture grade 1 green 10.5 (XL), 330mm long (approx.) Marigold