missing translation for 'onlineSavingsMsg'
Learn More

Faster™ GloveFAST Cyto Pharma Isolators

Product Code. 16113387
Click to view available options
Width (English):
98.82 in.
110.83 in.
122.8 in.
Width (English) Interior:
4 ft.
5 ft.
6 ft.
Includes:
2 Pass Boxes, 2 Gloves
2 Pass Boxes, 4 Gloves
This item is not returnable. View return policy

Product Code. 16113387

Brand: Faster™ F000206000FE

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GloveFAST Cyto Pharma isolators are suitable for the treatment of hazardous products and wherever product-personnel-environment cross-protection is required. Suitable for applications such as cytotoxic and CMR preparations, small production of cytotoxic or toxic drug, weighing, and dosing.

  • Two separated in/out transfer hatches for loading and unloading provided with 1 H14 HEPA/ULPA filter for inlet and a second H14 HEPA / ULPA filter for recirculation (electromagnetic interlocked doors system)
  • Low power consumption and noise level: GloveFAST Cyto Pharma offers high energy efficiency performance and silent operation that are achieved combining innovative solutions such as DC blowers, low pressure drop H14 HEPA/ULPA filters, and low power consumption LED light
  • LED lights system provides the highest lighting level (>1000 Lux) to the work area (dimmer is available as standard fitting to adjust light intensity)
  • Innovative system of third bank H 14 HEPA/ULPA filtration underneath work surface, filtering 100% of the air
    • This level of filtration is designed with an innovative Bag-Out System, allowing safe and easy replacement either for service engineers and environment. These filters have been designed to increase the space for operator's knees and comfort (>480 mm) as well as to satisfy the latest requirements of DIN 12980:2017.
  • 7° sloped front window provides optimum visibility of all objects placed in the workspace
    • Front laminated window 4+4 mm can be also opened and cleaned completely thanks to gas springs installed on the aluminum frame
  • Real Laminar AirFlow: Frontal screen 7° sloped as well as back side wall to convey in unidirectional pattern the air flow so that front and back panels are parallel one with the other and there is Laminar AirFlow in the whole working area
  • The new ECS™ microprocessor employs the latest innovative methods of integrated management of all principal functions of ventilation and filtration - self-regulating all the main filtration and ventilation system components, compensating for declining pressure drops and restoring power balance
    • Combining the use of DC motor-blowers and certified low pressure-drop filters, the new ECS™ controlling system optimize power consumption, reducing CO2 emissions into the environment
  • Dupont™ ALESTA™ Anti-bacterial 'Ag+ cations-based solution' coating prevents microbial contamination of surfaces, thereby inhibiting long term surface growth
  • High Level Lighting: Safety glass side-windows with the ideal positioning and sizing of the light-system provide the highest level of luminosity to the work area
  • Footswitch is supplied as standard in order to unlock the internal door of the chamber without removing hands from gloves
  • Automatic safety service connections for gas and vacuum include one (for size 312) or two (for size 315 and 318) electrical socket(s) fitted as standard in each size model
  • Work surface in stainless steel AISI 316 L, rear wall in stainless steel AISI 304 L

Specifications

Material Epoxy-coated Steel
Includes 2 Pass Boxes, 4 Gloves
Width (English) 122.8 in.
Width (English) Interior 6 ft.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.