missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GloLIVE Human/Mouse SSEA-1 NL557 Live Cell Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems NLLC2155R
This item is not returnable.
View return policy
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Mouse samples. It is validated for Immunocytochemistry.
Specifications
| SSEA-1 | |
| Monoclonal | |
| NorthernLights 557 (NL557) | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FUT4 | |
| F9 mouse teratocarcinoma stem cells. | |
| 0.5 mL | |
| Primary | |
| Detects human and mouse SSEA-1. | |
| Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. |
| Immunocytochemistry | |
| MC-480 | |
| Immunocytochemistry | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA1, SSEA-1, stage-specific embryonic antigen 1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 2526 | |
| Human, Mouse | |
| IgM |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur