missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLOD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16788-1-AP-150UL
This item is not returnable.
View return policy
Description
Germinal center kinases, such as TNIK, are characterized by an N-terminal kinase domain and a C-terminal GCK domain that serves a regulatory function.Specifications
| GLOD4 | |
| Polyclonal | |
| Unconjugated | |
| GLOD4 | |
| C17orf25, CGI 150, GLOD4, glyoxalase domain containing 4, HC71, My027 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 363644, 51031, 67201 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5I0D1, Q9CPV4, Q9HC38 | |
| GLOD4 | |
| GLOD4 Fusion Protein Ag10222 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction