missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Globo H Monoclonal Antibody (VK9), eBioscience™, Invitrogen™

Product Code. p-7218693
Change view
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17140752 2 mg 2mg
17150752 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17140752 Supplier Invitrogen™ Supplier No. 14970037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The VK9 MAb recognizes human Globo H, a hexasaccharide formed from the pentasaccharide precursor, SSEA3 (stage-specific embryonic antigen 3). It is a member of a family of antigenic carbohydrates expressed on the surface of cancer cells as glycolipids and possibly glycoproteins. Globo H is overexpressed on many carcinomas, such as breast, colon, ovarian, gastric, pancreatic, lung, and prostate. Expression is observed in more than half of all ductal, lobular, and tubular breast carcinomas, but not in non-epithelial breast tumors. Globo H is also expressed on cancer stem cells and human embryonic stem cells, although expression decreases when cells are differentiated toward both neural progenitor cells and to a lesser extent in endodermal lineage cells. Expression in normal tissues is limited to cells composing secretory borders. Globo H is considered a target for immunotherapy of many epithelial cancers.

Globo H is a hexasaccharide formed from the pentasaccharide precursor, SSEA3 (stage-specific embryonic antigen 3). It is a member of a family of antigenic carbohydrates expressed on the surface of cancer cells as glycolipids and possibly glycoproteins. Globo H is overexpressed on many carcinomas, such as breast, colon, ovarian, gastric, pancreatic, lung, and prostate. Expression is observed in more than half of all ductal, lobular, and tubular breast carcinomas, but not in non-epithelial breast tumors. Globo H is also expressed on cancer stem cells and human embryonic stem cells, although expression decreases when cells are differentiated toward both neural progenitor cells and to a lesser extent in endodermal lineage cells. Expression in normal tissues is limited to cells composing secretory borders. Globo H is considered a target for immunotherapy of many epithelial cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Globo H
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VK9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.09% sodium azide; pH 7.2
Gene Alias Globo H antigen; globoH; Globohexaosylceramide
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.