Learn More
Invitrogen™ GLO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579315
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human MCF-7 whole cell, human COLO320 whole cell, human 22RV1 whole cell, human HepG2 whole cell, human A431 whole cell, human U937 whole cell, human THP-1 whole cell, rat brain tissue, rat spleen tissue, rat thymus tissue, mouse brain tissue, mouse spleen tissue, mouse thymus tissue, mouse NIH/3T3 whole cell. IHC: human lung cancer tissue, human mammary cancer tissue, rat small intestine tissue, rat spleen tissue, mouse spleen tissue IP: U251 whole cell.
The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.
Specifications
| GLO1 | |
| Polyclonal | |
| Unconjugated | |
| GLO1 | |
| 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; aldoketomutase; AW550643; cb554; epididymis secretory protein Li 74; GLO1; Glo-1; Glo-1r; Glo1-r; Glo-1s; Glo1-s; GLOD1; glx I; GLY1; GLYI; glyoxalase 1; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; glyoxalase I; glyoxylase 1; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; lactoylglutathione lyase GLO1; methylglyoxalase; Qglo; S-D-lactoylglutathione methylglyoxal lyase; wu:fb82g09; YM9571.15C; YML004C; zgc:66035 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 109801, 2739, 294320 | |
| -20°C | |
| Lyophilized |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q04760, Q6P7Q4, Q9CPU0 | |
| GLO1 | |
| E. coli-derived human GLO1 recombinant protein (Position: A2-M184). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.