missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLO1 Monoclonal Antibody (Glo1a)
GREENER_CHOICE

Product Code. 12837772
Change view
Click to view available options
Quantity:
100 μg
20 μL
Unit Size:
100µg
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12837772 100 μg 100µg
12857772 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12837772 Supplier Invitrogen™ Supplier No. MA113029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GLO1 Monoclonal Antibody for Western Blot, IHC (P)

The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLO1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone Glo1a
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene GLO1
Gene Accession No. Q04760, Q6P7Q4, Q9CPU0
Gene Alias 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; aldoketomutase; AW550643; cb554; epididymis secretory protein Li 74; GLO1; Glo-1; Glo-1r; Glo1-r; Glo-1s; Glo1-s; GLOD1; glx I; GLY1; GLYI; glyoxalase 1; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; glyoxalase I; glyoxylase 1; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; lactoylglutathione lyase GLO1; methylglyoxalase; Qglo; S-D-lactoylglutathione methylglyoxal lyase; wu:fb82g09; YM9571.15C; YML004C; zgc:66035
Gene Symbols GLO1
Host Species Mouse
Immunogen Recombinant human GLOI-GST fusion protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109801, 2739, 294320
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.