missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLO1 Monoclonal Antibody (Glo1a)

Mouse Monoclonal Antibody
£188.00 - £477.00
Specifications
| Antigen | GLO1 |
|---|---|
| Clone | Glo1a |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12837772
![]() |
Invitrogen™
MA113029 |
100 μg |
£477.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
12857772
|
Invitrogen™
MA113029X |
20 μL |
£188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLO1 Monoclonal Antibody for Western Blot, IHC (P)
The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.Specifications
| GLO1 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Q04760, Q6P7Q4, Q9CPU0 | |
| 109801, 2739, 294320 | |
| Recombinant human GLOI-GST fusion protein. | |
| Antibody |
| Glo1a | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat, Monkey | |
| GLO1 | |
| 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; aldoketomutase; AW550643; cb554; epididymis secretory protein Li 74; GLO1; Glo-1; Glo-1r; Glo1-r; Glo-1s; Glo1-s; GLOD1; glx I; GLY1; GLYI; glyoxalase 1; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; glyoxalase I; glyoxylase 1; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; lactoylglutathione lyase GLO1; methylglyoxalase; Qglo; S-D-lactoylglutathione methylglyoxal lyase; wu:fb82g09; YM9571.15C; YML004C; zgc:66035 | |
| GLO1 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title