missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GlnRS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12645-1-AP-20UL
This item is not returnable.
View return policy
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. In metazoans, 9 aminoacyl-tRNA synthetases specific for glutamine (gln), glutamic acid (glu), and 7 other amino acids are associated within a multienzyme complex. Although present in eukaryotes, glutaminyl-tRNA synthetase (QARS) is absent from many prokaryotes, mitochondria, and chloroplasts, in which Gln-tRNA(Gln) is formed by transamidation of the misacylated Glu-tRNA(Gln). Glutaminyl-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family. Alternative splicing results in multiple transcript variants.Specifications
| GlnRS | |
| Polyclonal | |
| Unconjugated | |
| QARS | |
| GlnRS, Glutamine tRNA ligase, glutaminyl tRNA synthetase, PRO2195, QARS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 290868, 5859, 97541 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P47897, Q66H61, Q8BML9 | |
| Qars | |
| GlnRS Fusion Protein Ag3375 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction