missing translation for 'onlineSavingsMsg'
Learn More

GLN-1062, MedChemExpress

Product Code. 16440496
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16440496

Brand: Medchem Express HY101710

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Memogain is a pro-drug of galantamine, used for the treatment for Alzheimer’s disease.

Memogain is a pro-drug of galantamine, used for the treatment for Alzheimer’s disease.

Specifications

Chemical Name or Material GLN-1062
CAS 224169-27-1
Color White
Purity Grade Notes Research
Molecular Formula C24H25NO4
Quantity 20 mg
Solubility Information DMSO : 100 mg/mL (255.45 mM; Need ultrasonic)
SMILES CN1CC[C@]23C4=C(O[C@@]2([H])C[C@@H](OC(C5=CC=CC=C5)=O)C=C3)C(OC)=CC=C4C1
Molecular Weight (g/mol) 391.46
Formula Weight 391.46
Percent Purity 96.66%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.