missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLMN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35201-100ul
This item is not returnable.
View return policy
Description
GLMN Polyclonal antibody specifically detects GLMN in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| GLMN | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FAP, FAP48FAP68, FKBPAP, GLMLFK506-binding protein-associated protein, glomulin, glomulin, FKBP associated protein, GVM, GVMFKBP-associated protein, VMGLOMvenous malformation with glomus cells | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 405-594 of human GLMN (NP_444504.1).,, Sequence:, RCLLNTSNHSGVEAFIIQNIKNQIDMSLKRTRNNKWFTGPQLISLLDLVLFLPEGAETDLLQNSDRIMASLNLLRYLVIKDNENDNQTGLWTELGNIENNFLKPLHIGLNMSKAHYEAEIKNSQEAQKSKDLCSITVSGEEIPNMPPEMQLKVLHSALFTFDLIESVLARVEELIEIKTKSTSEENIGIK | |
| 100 μL | |
| Angiogenesis | |
| 11146 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction