missing translation for 'onlineSavingsMsg'
Learn More

Glisoxepid, TRC

Product Code. 30395098
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395098 100 mg
30395325 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395098 Supplier Toronto Research Chemicals Supplier No. TRCG410600100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25046-79-1
Molecular Formula C20 H27 N5 O5 S
Molecular Weight (g/mol) 449.52
Synonym 3-Isoxazolecarboxamide, N-[2-[4-[[[[(hexahydro-1H-azepin-1-yl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl-, Urea, 1-(hexahydro-1H-azepin-1-yl)-3-[[p-[2-(5-methyl-3-isoxazolecarboxamido)ethyl]phenyl]sulfonyl]- (8CI), N-[2-[4-[[[[(Hexahydro-1H-azepin-1-yl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl-3-isoxazolecarboxamide, 4-[4-[β-(5-Methylisoxazole-3-carboxamido)ethyl]phenylsulfonyl]-1,1-hexamethylenesemicarbazide, BS 4231, Glisepin, Glisoxepid, Glisoxepide, Pro-Diaban, Pro-Diaben, RP 22410
IUPAC Name N-[2-[4-(azepan-1-ylcarbamoylsulfamoyl)phenyl]ethyl]-5-methyl-1,2-oxazole-3-carboxamide
SMILES Cc1onc(c1)C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NN3CCCCCC3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H27N5O5S/c1-15-14-18(23-30-15)19(26)21-11-10-16-6-8-17(9-7-16)31(28,29)24-20(27)22-25-12-4-2-3-5-13-25/h6-9,14H,2-5,10-13H2,1H3,(H,21,26)(H2,22,24,27)
Formula Weight 449.1733
Chemical Name or Material Glisoxepide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.