missing translation for 'onlineSavingsMsg'
Learn More

GLIS3 Antibody (PCRP-GLIS3-1B11) - Azide and BSA Free, Novus Biologicals™

Product Code. 18321074 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321074 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321074 Supplier Novus Biologicals Supplier No. NBP314052

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GLIS3 Monoclonal antibody specifically detects GLIS3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLIS3
Applications Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array
Classification Monoclonal
Clone PCRP-GLIS3-1B11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 to 2 μg /million cells, ELISA, Immunoprecipitation 1 to 2 μg per 100-500 μg protein, Protein Array
Formulation 10mM PBS without Preservative
Gene Alias FLJ38999, FLJ90578, GLIS family zinc finger 3, GLI-similar 3, OK/KNS-cl.4, zinc finger protein 515, zinc finger protein GLIS3
Host Species Mouse
Immunogen Recombinant full-length human GLIS3 protein (Uniprot: Q8NEA6)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169792
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.