missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GliS2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£260.00 - £587.00
Specifications
| Antigen | GliS2 |
|---|---|
| Dilution | Western Blot 0.04 to 0.4 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18308282
|
Novus Biologicals
NBP3-17373-25UL |
25 μg |
£260.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18366492
|
Novus Biologicals
NBP3-17373-100UL |
100 μg |
£587.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GliS2 Polyclonal antibody specifically detects GliS2 in Human samples. It is validated for Western BlotSpecifications
| GliS2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, pH 7.2, 40% glycerol | |
| 84662 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04 to 0.4 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| GLIS family zinc finger 2, NKL, zinc finger protein GLIS2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VEGRFSAAPLVDLSLSPPSGLDSPNGSSSLSPERQGNGDLPPVPSASDFQPLRYLDGVPSS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title