Learn More
Invitrogen™ GLIS2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA540314
Product Code. 15594072
  • £456.00 / 100µL
Description
Peptide sequence: QDLVDHVNDY HVKPEKDAGY CCHWEGCARH GRGFNARYKM LIHIRTHTNE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.
Members of the Kruppel-like zinc finger protein family, such as GLIS2, function as activators and/or repressors of gene transcription.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GLIS2
Polyclonal
Unconjugated
GLIS2
Gli5; GLI-Kruppel family member GLI5; GLIS family zinc finger 2; GLIS2; GLI-similar 2; Klf16; Kruppel-like factor 16 (neuronal); Kruppel-like zinc finger protein GLIS2; nephrocystin-7; neuronal Krueppel-like protein; NKL; NPHP7; Zinc finger protein GLI5; zinc finger protein GLIS2
Rabbit
Affinity Chromatography
RUO
84662
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9BZE0
GLIS2
Synthetic peptide directed towards the N-terminal of human GLIS2 (aa 183-232).