missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLIS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572849
This item is not returnable.
View return policy
Description
GLIS2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Members of the Kruppel-like zinc finger protein family, such as GLIS2, function as activators and/or repressors of gene transcription.
Specifications
| GLIS2 | |
| Polyclonal | |
| Unconjugated | |
| GLIS2 | |
| Gli5; GLI-Kruppel family member GLI5; GLIS family zinc finger 2; GLIS2; GLI-similar 2; Klf16; Kruppel-like factor 16 (neuronal); Kruppel-like zinc finger protein GLIS2; nephrocystin-7; neuronal Krueppel-like protein; NKL; NPHP7; Zinc finger protein GLI5; zinc finger protein GLIS2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 302946, 83396, 84662 | |
| 4°C | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8VDL9, Q9BZE0 | |
| GLIS2 | |
| GLIS2 antibody for an 18 amino acid peptide near the center of human GLIS2. The immunogen is within amino acids 260 - 310 of GLIS2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction