missing translation for 'onlineSavingsMsg'
Learn More

Glis1 Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30734012 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734012 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734012 Supplier Novus Biologicals Supplier No. AF7618MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Glis1 Polyclonal antibody specifically detects Glis1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glis1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias FLJ36155, GLIS family zinc finger 1, GLI-similar 1, zinc finger protein GLIS1
Host Species Sheep
Immunogen E. coli-derived recombinant human Glis1, Met446-Thr620, Accession # Q5VTL4
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148979
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.