missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GLIPR1L2 Polyclonal antibody specifically detects GLIPR1L2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | GLIPR1L2 |
| Applications | Western Blot, Immunohistochemistry |
| Conjugate | mFluor Violet 450 SE |
| Gene Alias | GLI pathogenesis-related 1 like 2, GLIPR1-like protein 2, MGC39497 |
| Immunogen | GLIPR1L2 antibody was raised against an 15 amino acid peptide near the center of human GLIPR1L2. The immunogen is located within amino acids 120 - 170 of GLIPR1L2. Amino Acid Squence: RSWHAEKKMYNFENGS |
| Quantity | 0.1 mL |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?