missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLIPR1L2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82092H
This item is not returnable.
View return policy
Description
GLIPR1L2 Polyclonal antibody specifically detects GLIPR1L2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GLIPR1L2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| GLI pathogenesis-related 1 like 2, GLIPR1-like protein 2, MGC39497 | |
| GLIPR1L2 antibody was raised against an 15 amino acid peptide near the center of human GLIPR1L2. The immunogen is located within amino acids 120 - 170 of GLIPR1L2. Amino Acid Squence: RSWHAEKKMYNFENGS | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 144321 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction