missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLIPR1L2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-82092B
This item is not returnable.
View return policy
Description
GLIPR1L2 Polyclonal antibody specifically detects GLIPR1L2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| GLIPR1L2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| GLI pathogenesis-related 1 like 2, GLIPR1-like protein 2, MGC39497 | |
| GLIPR1L2 antibody was raised against an 15 amino acid peptide near the center of human GLIPR1L2. The immunogen is located within amino acids 120 - 170 of GLIPR1L2. Amino Acid Squence: RSWHAEKKMYNFENGS | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 144321 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction