missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLIPR1L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559865
This item is not returnable.
View return policy
Description
Immunogen sequence: SKIPSITDPH FIDNCIEAHN EWRGKVNPPA ADMKYMIWDK GLAKMAKAWA NQCKFEHNDC LDKSYKCYAA FEYVGENIWL GGIKS Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 47%.
Plays a role in the binding between sperm and oocytes. Component of epididymosomes, one type of membranous microvesicules which mediate the transfer of lipids and proteins to spermatozoa plasma membrane during epididymal maturation. Also component of the CD9-positive microvesicules found in the cauda region. [UniProt].
Specifications
| GLIPR1L1 | |
| Polyclonal | |
| Unconjugated | |
| GLIPR1L1 | |
| ALKN2972; GLI pathogenesis related 1 like 1; GLI pathogenesis-related 1 like 1; GLIPR1L1; GLIPR1-like protein 1; PRO7434; UNQ2972/PRO7434 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 256710 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UWM5 | |
| GLIPR1L1 | |
| Recombinant protein corresponding to Human GLIPR1L1. Recombinant protein control fragment (Product #RP-97830). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction